L(+)-Ascorbic acid 99.0-100.5%, crystalline powder USP, FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Ascorbic AcidL-Ascorbic acidAcidum ascorbicum(R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one

Management of Change (MOC) category = R

0936-08 BAKR0936-07 0936-07 BAKR0936-08
CAJT0936-8EA 2105.08 CAD
CAJT0936-8 CAJT0936-7 JT0936-7 JT0936-8
L(+)-Ascorbic acid 99.0-100.5%, crystalline powder USP, FCC, J.T.Baker®
L(+)-Ascorbic acid

  • Food GMP manufactured product

Formula: C₆H₈O₆
MW: 176.13 g/mol
Boiling Pt: 553 °C (1013 hPa)
Melting Pt: 190…192 °C
Density: 1.65 g/cm³ (20 °C)
MDL Number: MFCD00064328
CAS Number: 50-81-7
Merck Index: 13,00837
https://ca.vwr.com/stibo/web/std.lang.all/98/10/16009810.jpg https://ca.vwr.com/stibo/web/std.lang.all/05/91/16640591.jpg

Order Now

Specification Test Results

Meets F.C.C. Requirements
Food GMP Manufactured Product
GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (C₆H₈O₆) 99.0 - 100.5 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Specific Rotation [Å]²⁵ ^D (+) 20.5 - 21.5 Degree
USP - Residue on Ignition ≤0.1 %
FCC - Assay (C₆H₈O₆) 99.0 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Specific Rotation [Å]²⁵ ^D (+) 20.5 - 21.5 Degree
FCC - Residue on Ignition ≤0.1 %
Arsenic (As) ≤3 ppm
Mesh - Thru U.S. No. 20 Sieve ≥100 %
Mesh - Thru U.S. No. 100 Sieve ≤70 %
Mesh - Thru U.S. No. 200 Sieve ≤20 %
Residual Solvents - Only Class 2 Solvents (Methanol) and Class 3
Solvents are likely to be present. Residual Class 2 Solvents are below
Option 1 and the residual concentration of all Class 3 Solvents is
<0.5%.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR