Female Threaded Straight Couplings, PTFE
Supplier: MICROMOLD PRODUCTS, INC.
F-UN02
MFLX34030-68EA 346.02 CAD
MFLX34030-68
Female Threaded Straight Couplings, PTFE
Fittings and Connectors
Create a secure connection with threaded fittings.
  • Constructed of chemically inert PTFE
Chemically inert, PTFE offers outstanding resistance for any use, especially for critical high-purity applications. When subjected to higher pressure, PTFE as a material has a tendency to creep over time. As a result, these fittings work best at lower operating pressures (110 psi maximum operating pressure at ambient temperature for smaller sizes). All fittings are made from virgin PTFE material.