Female Threaded Cap Fittings, PTFE
Supplier: MICROMOLD PRODUCTS, INC.
F-CA06 F-CA04 F-CA02 F-CA01 F-CA08
MFLX34030-61EA 93.22 CAD
MFLX34030-61 MFLX34030-62 MFLX34030-65 MFLX34030-64 MFLX34030-66
Female Threaded Cap Fittings, PTFE
Fittings and Connectors
Chemically-inert PTFE handles most aggressive chemicals.
  • Constructed of durable PTFE
Chemically inert, PTFE offers outstanding resistance for any use, especially for critical high-purity applications. When subjected to higher pressure, PTFE as a material has a tendency to creep over time. As a result, these fittings work best at lower operating pressures (110 psi maximum operating pressure at ambient temperature for smaller sizes). All fittings are made from virgin PTFE material.