Bulkhead Fitting Connectors, PTFE
Supplier: MICROMOLD PRODUCTS, INC.
F-TA02 F-TA16 F-TA01 F-TA08 F-TA04
MFLX34031-14EA 579.86 CAD
MFLX34031-14 MFLX34031-16 MFLX34031-19 MFLX34031-11 MFLX34031-12
Bulkhead Fitting Connectors, PTFE
Fittings and Connectors
Suitable for use with flat-wall or most cylindrical, smooth-wall tanks.
  • Includes FEP encapsulated silicone O-ring
  • Body can be made longer to accommodate thicker tank walls
Constructed of durable PTFE, these Bulkhead Fitting Connectors are ideal for a wide range of applications.