Cole-Parmer® Essentials Pre-cleaned EPA Wide Mouth Bottles, Amber Glass, Antylia Scientific
Supplier: Antylia Scientific
99540-53 99540-55 99540-26 99540-51 99540-25 99540-57
76533-980CS 89.53 CAD
76533-980 76533-982 76533-976 76533-944 76533-978 76533-946
Cole-Parmer® Essentials Pre-cleaned EPA Wide Mouth Bottles, Amber Glass, Antylia Scientific
Bottles Glass Bottles
Certified clean for worry-free sampling
  • Includes certificate of conformance to EPA specifications
  • Leakproof PTFE-lined screw caps protect contents
  • Identify samples easier with included analysis labels (unattached)
  • Non-barcode label
  • Autoclavable
These pre-cleaned, wide-mouth bottles are processed and certified to meet the requirements of the current US EPA 'Specifications and Guidance for Contaminant-free Sample Containers' document for semi-volatile organics, pesticides, PCBs, metals, cyanide, fluoride, nitrate, and nitrite analyzes.