Affinity Chromatography Columns, Tagged Package, GST
Supplier: Cytiva
29-0588-04 29-0588-04
CA10498-070EA 369.89 CAD
10498-070 CA10498-070
Affinity Chromatography Columns, Tagged Package, GST
Chromatography Columns
The GST package is designed for easy purification of GST-tagged proteins.
The GSTrap™ 4B column (1×1 ml) has a high binding capacity for GST-tagged proteins, giving you higher yields. A HiTrap™ desalting column (1×5 ml) is provided for rapid buffer exchange.
Delivery information: The kit contains one of each GSTrap™ 4B (1×1 ml) and HiTrap™ Desalting (1×5 ml) columns.