Non-Marking ESD Heel Grounder, Transforming Technologies
Supplier: Transforming Technologies
HG1360NM-2
76474-660EA 21.61 CAD
76474-660
Non-Marking ESD Heel Grounder, Transforming Technologies
Grounders Heel and Toe Grounders
Non-marking stretch Hook and Loop heel grounders help keep controlled environments clean, and personnel grounded.
  • Made with ESD gray rubber that will not leave marks on floors
  • Stretchable hook and loop closure straps ensure a solid, comfortable fit
  • Reversible cup design allows for longer life
  • Made with a hi-vis blue strap that can be easily seen
The device is constructed from a non-marking and highly dissipative three-layer rubber. The reversible 1.5" wide rubber cup secures at the back and underneath the heel. The 7.5" length rubber provides enough cup volume to fit comfortably on those with larger shoe sizes. The 7" stretch hook and loop enclosure ensure a comfortable fit.
These heel grounders connect the person wearing them to ground via a proper floor mat or flooring material. Wearing the conductive ribbon inside the shoe or sock assures proper electrical contact with the user. A buried 1meg ohm resistor is standard. Heel grounders are worn on both feet to provide consistent grounding while in motion.
Certifications: Complies with ANSI ESD S2020.