BD Veritor™ Plus System, Antigen Testing for Rapid Detection of SARS-CoV-2
Supplier: BD Diagnostics
BD Veritor™
256066 256089
CA76463-386KT 1761.85 CAD
CA76463-386 CA75846-772
BD Veritor™ Plus System, Antigen Testing for Rapid Detection of SARS-CoV-2
Clinical Diagnostic Supplies Clinical Analysers
The BD Veritor™ Plus System is a handheld device that enables testing for Flu A+B, Group A Strep, and respiratory syncytial virus (RSV) assays.
  • Easy operation and 1-button functionality may potentially reduce manual test processing errors
  • Enables intuitive sample processing with prefilled, unitized tubes color-coded by reagent or assay type
  • Displays easy-to-read digital results for SARS-CoV-2 in 15 minutes
  • Records results on secured drive
  • Advanced particle technology enhances sensitivity by using a proprietary process to produce highly stable, modified colloidal metal particles, helping improve test performance
  • Adaptive read technology helps improve specificity to reduce false-positive results by compensating for background and nonspecific binding
One-button functionality and workflow flexibility allow the healthcare team to confidently use the device. In addition, the system’s fast and accurate results may help improve the patient experience. The BD Veritor™ Plus System streamlines the point-of-care testing experience.