UltraLabel PRO™ Cleanroom Printers, UltraTape
Supplier: ULTRATAPE INDUSTRIES
ULP300GY-D ULP300GY-S-N
89179-504EA 3912.13 CAD
89179-504 89179-502
UltraLabel PRO™ Cleanroom Printers, UltraTape
Printers Document Printers
Complete print-on-demand solutions that can be customized to specific application requirements.
  • Highly versatile cleanroom printer
  • Handles most labeling and sign making tasks
  • Easy to set up and ready to go right out of the box
  • Fast, high resolution 300 dpi printing
  • Versatility of direct thermal or thermal transfer printing
The UltraLabel PRO™ is a desktop 'thermal transfer/direct thermal' cleanroom label and sign printer. It handles a wide variety of jobs ranging from printing small die-cut cleanroom labels and shrink tube wire labels, to making reflective safety signs. From inventory and product labels to pipe markers or hazardous materials labels, the UltraLabel PRO™ provides excellent print quality and saves money by allowing in-house cleanroom labeling.