Papanicolaou's Orange G solution (OG 6), HARLECO® stain for cytology, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
7052X-75
7052X-86
7052X-86
7052X-85
7052X-75
7052X-85
CA15204-276EA
297.82
CAD
CA15204-276
15204-206
15204-208
CA-15204-206L
CA15204-208
15204-276
Papanicolaou's Orange G solution (OG 6), HARLECO® stain for cytology, Sigma-Aldrich®
Papanicolaou's Orange G solution (OG 6)
Cytoplasmic counterstain for Pap staining technique.
Prepared from dyes certified by the Biological Stain Commission (BSC)