Kimble® KONTES® DISPOSAFLEX® Disposable Columns, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
420160-0000
KT420160-0000PK 438.83 CAD
KT420160-0000
Kimble® KONTES® DISPOSAFLEX® Disposable Columns, DWK Life Sciences
Chromatography Columns
An inexpensive, multi-purpose column designed to satisfy a wide range of analytical separations using aqueous buffers. A typical application of the Disposaflex® column is the extraction of the cyclic nucleotides cAMP and cGMP from sample preparations using AG1-X8.
  • The disposable feature eliminates the possibility of cross-contamination when using labeled compounds
  • The 8 mm inner diameter of the polypropylene column is uniform, assuring reproducibility
  • The length of the DISPOSAFLEX® column can easily be cut shorter
  • Polyethylene bed supports and on-off valves are available as accessories
The lower column fitting with a Luer outlet is molded polypropylene with a specially designed recess to securely hold a porous polyethylene bed support or a glass wool plug.
Delivery information: Supplied unassembled with a column, polypropylene reservoir and lower column fitting.