DuPont™ ProClean® 2 Sterile Sleeves
Supplier: DuPont
GB500SBU000S
76290-262CS 873.44 CAD
76290-262
DuPont™ ProClean® 2 Sterile Sleeves
DuPont ProClean® 2 sterile sleeves are made from poly/cellulose.
  • Sterile
  • Elastic at both ends
  • Low lint generation
  • Universal fit
Sterile sleeves are individually packaged, easy to open, and easy to store. When the sleeve of your sterile gown is contaminated, or you have a strikethrough, you need to do something fast. Protect yourself with this sterile sleeve. Elastic at both ends of the sleeve makes it easy to put on right over your sterile gown, so you don't need to regown.
Sleeves feature elastic at both ends for a universal fit, with a 3" wrist opening and a 5 1/2" bicep opening.
Certifications: EO Sterilization with 10-6. Thickness meets ASTM D3776.
Packaging: Sterile sleeves are individually packaged with 100 sleeves per case.