Anti-IL-16 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P06-50UG 500-P06-100UG 500-P06-1MG
10775-006EA 536.88 CAD
10775-006 10775-008 10781-464
Anti-IL-16 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IL-16. Anti­Human IL-16­specific antibody was purified by affinity chromatography employing an immobilized Human IL-16 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR