Anti-VWF Mouse Monoclonal Antibody (CF405S) [clone: VWF635]
Supplier: Biotium
BNC040635-100 BNC040635-500
75979-716EA 325.12 CAD
75979-716 75979-718
Anti-VWF Mouse Monoclonal Antibody (CF405S) [clone: VWF635]
Antibodies
von Willebrand Factor (vWF) is a multimeric glycoprotein that is found in endothelial cells, plasma and platelets. It acts as a carrier protein for Factor VIII and promotes platelet adhesion and aggregation. vWF undergoes a variety of posttranslational modifications that influence the affinity and availability for Factor VIII, including cleavage of the propeptide and formation of N-terminal disulfide bonds. This antibody helps to establish the endothelial nature of some lesions of disputed histogenesis, e.g. Kaposi's sarcoma and cardiac myxoma. It is widely used for differentiating vascularMore Product Information