L(+)-Potassium sodium tartrate tetrahydrate ≥99.0%, powder FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrateRochelle saltL(+)-Tartaric acid potassium sodium salt tetrahydratePotassium sodium tartrate tetrahydratePotassium sodium L(+)-tartrate tetrahydrate(+)-Potassium sodium L-tartrate tetrahydrateSodium potassium tartratePotassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate(R,R)-(+)-Potassium sodium tartrate tetrahydrate

P1402-12KG P1402-500GM P1402-2.5KG
75811-938EA 3270.09 CAD
95035-210 95035-208 75811-938
L(+)-Potassium sodium tartrate tetrahydrate ≥99.0%, powder FCC
L(+)-Potassium sodium tartrate tetrahydrate

Sodium Potassium Tartrate, Tetrahydrate, Powder, FCC is used as a sequestrant and stabilizer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: KOOC(CHOH)₂COONa·4H₂O
MW: 282.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150989
CAS Number: 6381-59-5
https://ca.vwr.com/stibo/web/std.lang.all/98/18/2299818.jpg

Order Now

Specification Test Results

Assay 99.0-102.0 %
Alkalinity To Pass Test
Water 21.0- 26.0 %
Lead (Pb) max. 2 mg/kg
Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR