Personal Air Cooler, Allegro
Supplier: Allegro
9991
57006-058EA 703.97 CAD
57006-058
Personal Air Cooler, Allegro
Respirators Respiratory Systems
Vortex Tubes are an effective, low-cost solution to respiratory cooling needs.
  • Uses filtered compressed air
  • Provides continuous cool air delivery
  • Easy temperature adjustment even with gloved hands
  • No moving parts
  • Control valve allows for temperature adjustment
  • Will cool compressed air by 25° F
Delivering constant and adjustable cooling or heating, the Vortex Vest is especially popular for workers in confined spaces as well as foundries, steel mills, asbestos abatement and paint baking operations
With no moving parts, the vortex tube forces a simple heat exchange to separate compressed air into hot and cold airstreams.
Body: Aluminum
Fitting: Hansen 1/4” (.63cm) plug (series 1000) Stainless Steel
Belt: Nylon
Please note: Requires an air supply pressure range from 50-115 psi.