HP35 High-Profile Blades, Epredia™, Richard-Allan Scientific
Supplier: Richard-Allan Scientific
Epredia™
3150734 3150734
CA14011-014PK 947.64 CAD
CA14011-014 89238-786
HP35 High-Profile Blades, Epredia™, Richard-Allan Scientific
Microtomes and Microtome Assemblies Microtome Blades and Handles
Use Epredia™ HP35 Disposable Microtome Blades for routine applications with a high proportion of harder tissue.
  • High profile
  • Increased durability