Oxalic acid dihydrate, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Ethanedioic acid dihydrate

M-Clarity™ quality level = MQ200

8.18242.9025
EM8.18242.9025EA 634.69 CAD
EM8.18242.9025
Oxalic acid dihydrate, Sigma-Aldrich®
Oxalic acid dihydrate
Formula: HO₂CCO₂H·2H₂O
MW: 126.07 g/mol
Boiling Pt: 149…160 °C (1013 hPa)
Melting Pt: 101 °C
Density: 1.65 g/cm³ (20 °C)
MDL Number: MFCD00149102
CAS Number: 6153-56-6
UN: 3261
ADR: 8,III
https://ca.vwr.com/stibo/web/std.lang.all/72/16/2297216.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR