Disposable Micropipettes, Analtech
Supplier: Miles Scientific
20-13
47746-458PK 102.95 CAD
47746-458
Disposable Micropipettes, Analtech
Pipettes Microcapillaries
These thin-bore capillary tubes are ideal for applying TLC samples when high accuracy and precision are not required.
Sample volume is estimated visually.