TX-100 Swinging Bucket Clinical Rotor, Thermo Scientific
Supplier: Thermo Scientific
75005705
89423-778EA 5057.44 CAD
89423-778
TX-100 Swinging Bucket Clinical Rotor, Thermo Scientific
Centrifuges Rotors
Efficiently process up to 16 x 5/7 mL blood collection tubes per run.
Ordering information: Includes unsealed carriers. Adapters are sold separately. Contact your sales representative for more information.