Anti-CD154 Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: 24-31]

Supplier: Biolegend
310837 310838
CA10758-022EA 360.65 CAD
CA10758-022 CA10758-024
Anti-CD154 Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: 24-31]
Antibodies
Brilliant Violet 711™ anti-human CD154 [24-31]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD154
Clonality: Monoclonal
Clone: 24-31
Conjugation: Brilliant Violet® 711
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR