SP Wilmad-LabGlass Schlenk Vacuum Sample Tubes, SP Industries
Supplier: SP Industries
LG-10800-102 LG-10800-100
14204-310EA 1003.03 CAD
14204-310 14204-190
SP Wilmad-LabGlass Schlenk Vacuum Sample Tubes, SP Industries
Tubes Test Tubes
These borosilicate glass Schlenk sample tubes incorporate a grease-free high vacuum 0 to 4 mm valve and are suited for pressure/vacuum applications.
  • NOTE: 25 ml model has a round bottom
Ideal for reaction and storage of air-sensitive compounds. Sidearm can be connected to inert atmosphere/vacuum source or a manifold.