Anti-IgG Goat Polyclonal Antibody (Cy5®)

Supplier: Cytiva
PA45011 PA45011
CA95040-050LEA 483.47 CAD
95040-050 CA95040-050L
Anti-IgG Goat Polyclonal Antibody (Cy5®)
Antibodies
ECL Plex Cy-conjugated Antibodies are part of an optimized system for quantitative analysis using fluorescent western blotting.

  • For 1000 cm² membrane area

Optimized system for quantitative analysis using fluorescent Western blotting. ECL Plex Western Blotting System is optimized for single protein detection as well as multiplex protein detection using ECL Plex CyDye-Conjugated Secondary Antibodies and Hybond-LFP (low-fluorescent PVDF membrane) or Hybond ECL (nitrocellulose membrane). The optimized antibodies show high sensitivity, a very good dynamic range and low or no cross-reactivity. CyDye technology is dependable, reliable, and ensures accurate quantitation. The system is optimized on the Typhoon 9410 Scanner but is compatible with many fluorescent laser scanners and CCD cameras.

ECL Plex Western Blotting System is optimized for single protein detection as well as multiplex protein detection using ECL Plex CyDye-Conjugated Secondary Antibodies and Hybond-LFP (low-fluorescent PVDF membrane) or Hybond ECL (nitrocellulose membrane). The optimized antibodies show high sensitivity, a very good dynamic range and low or no cross-reactivity. CyDye technology is dependable, reliable, and ensures accurate quantitation. The system is optimized on the Typhoon 9410 Scanner but is compatible with many fluorescent laser scanners and CCD cameras.

Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR