7,7-Dichlorobicyclo[4.1.0]heptane ≥98.0% (by GC)

Supplier: TCI America

D4314-25G D4314-5G
TCD4314-25GEA 317.58 CAD
TCD4314-25G TCD4314-5G
7,7-Dichlorobicyclo[4.1.0]heptane ≥98.0% (by GC)
7,7-Dichlorobicyclo[4.1.0]heptane

CAS Number: 823-69-8
MDL Number: MFCD00466186
Molecular Formula: C7H10Cl2
Molecular Weight: 165.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Specific Gravity (20/20): 1.22

Formula: C₇H₁₀Cl₂
Boiling Pt: 80 °C
Density: 1.22 (20 °C)
MDL Number: MFCD00466186
CAS Number: 823-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4987f9e83c113b894c4d826dd4df3c0a4b9cf295&vid=4987f9e83c113b894c4d826dd4df3c0a4b9cf295&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR