Anti-HMOX1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-895-J SPA-895J
95046-310EA 3239 CAD
95046-310 CA95046-310
Anti-HMOX1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HMOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Sheep, Rabbit, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR