1-Pentanesulfonic acid sodium salt, Supelco®

Supplier: MilliporeSigma

Synonyms: Sodium pentane-1-sulfonateSodium pentanesulphonate, 98%Pentane-1-sulfonic acid sodium saltSodium pentane-1-sulphonateSodium pentanesulfonateSodium 1-pentanesulfonateSodium pentanesulfonate, 98%Pentane-1-sulphonic acid sodium saltSodium pentanesulphonateSodium 1-pentanesulphonate

M-Clarity™ quality level = MQ100

1.18304.0025 1.18304.0025
CA1.18304.0025CS 62174.47 CAD
EM1.18304.0025 CA1.18304.0025
1-Pentanesulfonic acid sodium salt, Supelco®
1-Pentanesulphonic acid sodium salt

Pentane-1-sulfonic acid sodium salt for ion pair chromatography LiChropur®.

Formula: C₅H₁₁O₃SNa
MW: 174.2 g/mol
MDL Number: MFCD00007541
CAS Number: 22767-49-3
https://ca.vwr.com/stibo/web/std.lang.all/09/27/18570927.jpg

Order Now

Specification Test Results

Hill Formula C₅H₁₁NaO₃S
Molar Mass 174.20 g/mol
pH value 5.5 - 7.5 (100 g/l, H₂O, 20 °C)
Bulk density 390 kg/m³
Solubility (20 °C) soluble
Storage class 10 - 13 Other liquids and solids
WGK WGK 3 highly water endangering

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR