Melanocyte Growth Medium M2, PromoCell
Supplier: PromoCell
C-24400 C-24405 C-24305 C-24300
10172-088EA 764.86 CAD
10172-088 10172-090 10172-092 10172-086
Melanocyte Growth Medium M2, PromoCell
Cell Culture Media
The unique PromoCell Melanocyte Growth Medium 2 is a serum-free and PMA (Phorbol Myristate Acetate) free medium optimized for the cultivation of human melanocytes
Recommended for Normal Human Epidermal Melanocytes (NHEM), juvenile foreskin, cultured in M2 Medium, Normal Human Epidermal Melanocytes (NHEM), adult donor, cultured in M2 Medium
It is optimized for primary human cells but can also be used for equine, porcine, murine, and rat melanocytes. It is also suitable for human pigment epithelial cells, neuronal cells and melanoma cells.
Ordering information: PromoCell Melanocyte Growth Medium M2 is available as Medium (Ready-touse) which consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. Adding the SupplementMix to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) and the SupplementMix can also be purchased separately.