Renal Epithelial Cell Growth Medium, PromoCell
Supplier: PromoCell
C-26235 C-26230 C-26130 C-26030
10172-322EA 213.08 CAD
10172-322 10172-314 10172-324 10172-318
Renal Epithelial Cell Growth Medium, PromoCell
Cell Culture Media
The PromoCell Renal Epithelial Cell Growth Medium is a serum-free medium for the cultivation of epithelial cells from the kidneys
Recommended for Human Renal Epithelial Cells (HREpC), Human Renal Cortical Epithelial Cells (HRCEpC).
The PromoCell Renal Epithelial Cell Growth Medium 2 is an optimized low-serum (5% V/V) medium with epinephrine. The combination of low serum and selected growth factors ensures successful growth in vitro and makes this medium the first choice for renal epithelial cells. The media are optimized for primary human cells but can also be used for porcine renal epithelial cells
Ordering information: PromoCell Renal Epithelial Cell Media are available as Medium (Ready-to-use) or as Medium Kit. The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as SupplementMix and SupplementPacks can also be purchased separately.