Magnesium sulfate

Supplier: G-Biosciences

Synonyms: MgSO4Magnesium sulfate anhydrousMagnesium sulphate anhydrous

M006 M006
CA89233-950EA 231.61 CAD
CA89233-950 89233-950
Magnesium sulfate
Magnesium sulphate
Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1127 °C
Density: 2.65 g/cm³ (20 °C)
MDL Number: MFCD00011110
CAS Number: 7487-88-9
Merck Index: 12,05731
https://ca.vwr.com/stibo/web/std.lang.all/06/17/16640617.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR