Stirrer Blade, Polypropylene, Dynalon
Supplier: Dynalon
212615
71070-132CS 696.94 CAD
71070-132
Stirrer Blade, Polypropylene, Dynalon
Stirrers Stirring Blades
Centrifugal polypropylene stirrer blades are 100mm in width (fully extended)
The blades are made of chemically resistant polypropylene and are fixed to a PVC-covered steel steel rod with a diameter of 8x350mm (approx. ⁵⁄₁₆ x 13¾") long. Clockwise motion directs flow upward.
Ordering information: Elasticity of tubing will influence choice of size. Dimensions listed are nominal.