Tin(IV) oxide ≥99.0% (RFA), EMSURE®, extra pure for analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: Tin(iv)oxide, Stannic Oxide, Tin dioxide

1.07818.0250 1.07818.9025 1.07818.0250 1.07818.9025
EM1.07818.0250CS 4455.6 CAD
EM1.07818.0250 EM1.07818.1000 EM1.07818.5000 EM1.07818.9025 CA1.07818.0250 CA1.07818.1000 CA1.07818.5000 CA1.07818.9025
Tin(IV) oxide ≥99.0% (RFA), EMSURE®, extra pure for analysis, Supelco®
Tin (IV) oxide
Formula: O₂Sn
Boiling Pt: 1800…1900 °C (subl.)
Melting Pt: 1630 °C
Density: 6.95 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011244
CAS Number: 18282-10-5

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 1630 °C
Molar Mass 150.69 g/mol
Bulk Density 500 - 600 kg/m3
Boiling Point 1800 - 1900 °C (sublimed)
Density 6.95 g/cm3 (20 °C)
pH 4 - 5 (50 g/l, H2O, 20 °C) (slurry)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR