Orion™ ORP Standards for Redox/ORP Electrodes, Thermo Scientific
Supplier: Thermo Scientific
967961 967901 967901 967961
CA46610-337PK 746.19 CAD
46610-336 CA46610-337 46610-338 CA46610-338
Orion™ ORP Standards for Redox/ORP Electrodes, Thermo Scientific
Standards Redox/ORP Standards
Used as relative mV standard to calibrate the offset of sample.
Stable for up to a year if unopened and can be used directly from the bottle with no mixing or diluting.
ORP standard does not contain hazardous chemicals.