Amersham™ CyDye™ Fluorescent Nucleotides
Supplier: Cytiva
CY™
PA53021 PA53021 25-8010-86 PA53022 25-8010-87 PA55321 PA55322 PA53521 25-8010-87 PA55021 PA55026 PA53031 PA55026 PA53026 PA55021 PA55321 PA55022 PA53026 25-8010-86 PA55022 PA53031 PA55322 PA53521 PA53022
CA95040-150LEA 8359.23 CAD
95040-148 95040-154 95040-142 95040-140 95040-158 95040-146 95040-168 95040-156 95040-144 95040-166 CA95040-150L CA95040-140L CA95040-142L 95040-150 95040-160 CA95040-160L CA95040-158L CA95040-148L CA95040-154L CA95040-144L CA95040-166L CA95040-156L CA95040-168L CA95040-146L
Amersham™ CyDye™ Fluorescent Nucleotides
Protein Linking and Labeling Systems
Fluorescent nucleotides for labeling DNA and RNA probes, especially during multi-colour analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping.
  • High sensitivity and low non specific binding
  • High photostability, high water solubility
  • pH insensitive
CyDye™ labelled nucleotides are available in a range of bright and intense colours with narrow emission bands for multiplexing with a single sample.