Lactobacilli MRS Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
G117 G197 U476
CA89407-144PK 50.92 CAD
CA89407-144 CA89407-196 CA89407-214 76299-188
Lactobacilli MRS Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Lactobacilli MRS Agar is recommended for the isolation, enumeration and cultivation of Lactobacillus spp.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature 2-8°C