Ac-VDVAD-CHO
Supplier: Enzo Life Sciences
260-058-M001 260-058-M005
89146-716EA 178.54 CAD
89146-716 89146-718
Ac-VDVAD-CHO
Proteins and Peptides
Caspase-2 inhibitor
Potent, reversible aldehyde inhibitor of caspase-2 (Ki=3.5nM). Preferred cleavage sequence for caspase-2.