Anti-PKC Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-PK003-E KAP-PK003E
CA200062-116EA 557.99 CAD
200062-116 CA200062-116
Anti-PKC Rabbit Polyclonal Antibody
Antibodies
Protein Kinase C (PKC) is a large superfamily of serine/threonine kinases that mediate essential cellular signals required for activation, proliferation, differentiation and survival. There are at least ten PKC isotypes that are closely related in structure but that have distinct patterns of tissue distribution and function. The PKC isotypes can be subdivided into three classes based on primary structure and biochemical properties. These are: classical or conventional PKC isotypes (cPKC), novel PKC isotypes (nPKC) and atypical PKC isotypes (aPKC).

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, dog, chicken, guinea pig, monkey, pig, rabbit and sheep PKC. Detects a band of ~67-83kDa by Western blot.

Type: Primary
Antigen: PKC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR