L(+)-Cysteine hydrochloride monohydrate

Supplier: BACHEM AMERICAS INC

Synonyms: (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate(R)-(+)-Cysteine hydrochloride monohydrateL-Cysteine HCI MonohydrateCysteine Hydrochloride monohydrateH-Cys-OH.HCl.H2OCysteine Hydrochloride, Monohydrate

4030461.0100
E-1755.0100BAEA 528.27 CAD
E-1755.0100BA
L(+)-Cysteine hydrochloride monohydrate
L(+)-Cysteine hydrochloride monohydrate

Sequence: H-Cys-OH · HCl · H₂O


Due to its high reactivity towards formaldehyde, cysteine should be added as scavenger when cleaving His(Bom)-containing peptides, especially in case of an N-terminal Cys. CAS Number (net): 52-90-4.

Formula: HSCH₂CH(NH₂)COOH·HCl·H₂O
MW: 175.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065606
CAS Number: 7048-04-6
https://ca.vwr.com/stibo/web/std.lang.all/58/37/2345837.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR