4-Aminoantipyrine ≥99.0%

Supplier: TCI America
Warning

Synonyms: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone, 4-Aminophenazone

A0256-500G A0256-25G
TCA0256-500GEA 965.38 CAD
TCA0256-500G TCA0256-025G
4-Aminoantipyrine ≥99.0%
4-Aminoantipyrine

CAS Number: 83-07-8
MDL Number: MFCD00003145
Molecular Formula: C11H13N3O
Molecular Weight: 203.25
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 109


Other Product Names / Synonyms: 4-Aminophenazone, 4-AA

Formula: C₁₁H₁₃N₃O
Melting Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00003145
CAS Number: 83-07-8
EINECS: 201-452-3
Merck Index: 13,00596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a94b41fb0d5a1f409308b793ef1610e86cfe4a1&vid=3a94b41fb0d5a1f409308b793ef1610e86cfe4a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR