Septum Puller, Restek
Supplier: Restek
20117
RK20117EA 53.87 CAD
RK20117
Septum Puller, Restek
Puller and Inserter Tools
Septum pullers lend an enhanced level of comfort in easily removing septa and O-rings from GC systems.
  • Effective at removing septa
  • Easily clear debris with fine pointed end
Use hooked end for removing septa and O-rings. The pointed end works well for removing stuck ferrules or debris.