Berberine chloride

Supplier: Thermo Scientific Chemicals

Synonyms: 9,10-Dimethoxy-5,6-dihydro-[1,3]dioxolo[4,5-g]isoquinolino[3,2-a]isoquinolin-7-ium chloride

J62311-06
CAAAJ62311-06EA 108.07 CAD
CAAAJ62311-06
Berberine chloride
Berberine chloride

Apoptosis inhibitor. Increases P-glycoprotein expression in hepatoma cells. Stain for heparin in mast cells


Powder


Applications: Apoptosis inhibitor. Increases P-glycoprotein expression in hepatoma cells. Stain for heparin in mast cells

Formula: C₂₀H₁₈ClNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00011939
CAS Number: 633-65-8
EINECS: 211-195-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR