D(-)-Lactic acid

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (R)-2-Hydroxypropanoic acid, D-Lactic Acid, (R)-(-)-2-Hydroxypropionic acid, (R)-(-)-Lactic acid, (R)-(-)-2-Hydroxypropanoic acid

J61871-03 J61871-MD
CAAAJ61871-03EA 1923.89 CAD
CAAAJ61871-03 CAAAJ61871-MD
D(-)-Lactic acid
D(-)-Lactic acid

Useful chiral synthon, building block for depsipeptides


Powder


Useful chiral synthon, building block for depsipeptides

Formula: C₃H₆O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00068313
CAS Number: 10326-41-7
EINECS: 233-713-2
Merck Index: 14,5335

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR