4-Bromobenzhydrazide 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromobenzene-1-carbohydrazide, 4-Bromobenzoic acid hydrazide, 4-Bromobenzohydrazide, 4-Bromobenzoic hydrazide

A18918-06 A18918-14
CAAAAA18918-22EA 1291.59 CAD
CAAAAA18918-22 CAAAA18918-06 CAAAA18918-14
4-Bromobenzhydrazide 98+%
4-Bromobenzhydrazide

Purity: 98+% CAS No.: 5933-32-4 Molecular Formula: C7H7BrN2O Formula Weight: 215.05 Melting Point: 167-169° Beilstein Reference: 777006

Formula: C₇H₇BrN₂O
Melting Pt: 167…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00007602
CAS Number: 5933-32-4
EINECS: 227-681-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR