Potassium ethanolate 15% (w/v) in ethanol , 99+% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium ethylate, Potassium ethoxide

39161-AC
CAAA39161-ACEA 150.26 CAD
CAAA39161-AC
Potassium ethanolate 15% (w/v) in ethanol , 99+% (metals basis)
Potassium ethanolate

Purity: 99+% CAS No.: 917-58-8 Molecular Formula: C2H5KO Formula Weight: 84.16 Physical Form: Liquid Sensitivity: Air and Moisture Sensitive Appearance: Colorless to pale yellow Odor: Alcohol-like UN No.: 3274 MDL No.: MFCD00054277

Formula: C₂H₅KO
Storage Temperature: Ambient
MDL Number: MFCD00054277
CAS Number: 917-58-8
EINECS: 213-029-0
UN: 3274
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR