CarePac® Test Weight Sets, METTLER TOLEDO®
Supplier: Mettler Toledo
11123100 11123102 11123103 11123107 11123108 11123109 11123110 11123144 11123145 11123146 11123147 11123148 11123149 11123150 11123151 11123152 11123153 11123154 11123155 11123156 11123157 11123158 11123159 11123008 11123009 11123010 11123044 11123045 11123046 11123047 11123048 11123049 11123050 11123051 11123052 11123053 11123054 11123055 11123056 11123057 11123058 11123059 11123111 11123011 11123101 11123105 11123104 11123004 11123106
CA97007-250EA 917.85 CAD
CA97007-250 CA97007-254 CA97007-256 CA97007-258 CA97007-260 CA97007-262 CA97007-264 CA97007-266 CA97007-268 CA97007-270 CA97007-272 CA97007-274 CA97007-276 CA97007-278 CA97007-280 CA97007-282 CA97007-284 CA97007-286 CA97007-288 CA97007-290 CA97007-292 CA97007-294 CA97007-296 CA97007-308 CA97007-310 CA97007-312 CA97007-314 CA97007-316 CA97007-318 CA97007-320 CA97007-322 CA97007-324 CA97007-326 CA97007-328 CA97007-330 CA97007-332 CA97007-334 CA97007-336 CA97007-338 CA97007-340 CA97007-342 CA97007-344 CA97027-716 CA97027-718 CA97007-252 97019-006 97019-008 97019-014 97019-004
CarePac® Test Weight Sets, METTLER TOLEDO®
Weights Calibration Weights
These sets enable routine testing of balances by using two weights at maximum and minimum loads
  • Routine testing of balances
  • Multiple configurations
Weights are available in both ASTM and OIML configurations and are specified to validate process tolerances up to 0.03%. Sets include ergonomic tweezers and cleaning cloths. Customized third weights are available for customer-specific testing, such as minimum weight determination. Specially-designed cases house both types of weights and all equipment. Additional space is provided to accommodate a third weight.