Guanidinium hydrochloride for molecular biology, Millipore®

Supplier: MilliporeSigma

369080-100GM 369080-1KG
CA80502-626EA 434.19 CAD
CA80502-626 CA80502-628
Guanidinium hydrochloride for molecular biology, Millipore®
Guanidine hydrochloride
Formula: NH₂C(=NH)NH₂·HCl
MDL Number: MFCD00013026
CAS Number: 50-01-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Molecular Weight 95.5

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR