Rotors for Centrifuge 5702/5702 R/5702 RH
Supplier: Eppendorf
022639501 022639404 022639421 022639471 022639048 022639064
CA47743-474EA 746.37 CAD
CA47743-474 CA47743-480 CA47743-482 CA80086-006 CA47743-508 CA47743-528
Rotors for Centrifuge 5702/5702 R/5702 RH
Centrifuges Rotors
Eppendorf rotors for centrifuge 5702 series - precision in every spin. Tailor your centrifugation with varied capacities and speeds. Accessorize with adaptable buckets for versatile sample processing. Ideal for low-speed applications, ensuring efficiency and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
Elevate your low-speed centrifugation with EppendorfMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification Renewable Energy Certificate, ISO 14001