5-Nitropicolinic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning

H64499-03 H64499-06
AAH64499-03EA 57.35 CAD
AAH64499-03 AAH64499-06 AAH64499-14
5-Nitropicolinic acid 97+%
5-Nitropicolinic acid

5-Nitropyridine-2-carboxylic acid, 97+%


Purity: 97+% CAS No.: 30651-24-2 Molecular Formula: C6H4N2O4 Formula Weight: 168.11 Physical Form: Solid Melting Point: 208-212° Beilstein Reference: 139209 MDL No.: MFCD04114182

Formula: C₆H₄N₂O₄
Melting Pt: 208…212 °C
Storage Temperature: Ambient
MDL Number: MFCD04114182
CAS Number: 30651-24-2

Order Now

Specification Test Results

Assay 97%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR