Human Recombinant Omentin 1
Supplier: Enzo Life Sciences
522-108-C010
89151-888EA 821.6 CAD
89151-888
Human Recombinant Omentin 1
Proteins and Peptides
Produced in HEK 293 cells. Human omentin 1 (intelectin-1) (aa 17-313) is fused at the N-terminus to a FLAG®-tag.
Has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes. Increases AKT phosphorylation in the absence and presence of insulin. May play a role in the defense system against microorganisms. May specifically recognize carbohydrate chains of pathogens and bacterial components containing galactofuranosyl residues, in a calcium-dependent manner. May be involved in iron metabolism.