BAPTA tetracesium salt ≥95% (by TLC) ion chelator, for calcium determination

Supplier: Biotium

Warning

50001
89139-156EA 801.96 CAD
89139-156
BAPTA tetracesium salt ≥95% (by TLC) ion chelator, for calcium determination
BAPTA tetracesium salt

BAPTA and its derivatives are calcium chelators that are commonly used to form calcium buffers with well-defined calcium concentrations.


  • Kd (no Mg2 ): 0.59 uM, Kd (1 mM Mg2 ): 0.70 uM
  • White solid soluble in water
  • Store at 4°C


By injecting the chelators into cells or by incubating cells with the AM ester form of the chelators, one can control the cytosolic calcium concentration, an important means to study the roles of calcium. Key advantages of these calcium chelators include relative insensitivity toward intracellular pH change and fast release of calcium. Biotium offers several BAPTA chelators with calcium dissociation constants covering the biologically significant range from 10-7 to 10-2 M.

Formula: C₂₃H₂₄Cs₄N₂O₁₀
CAS Number: 480436-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR