Orion™ AquaMate Vis and UV-Vis Spectrophotometers, Thermo Scientific
Supplier: Thermo Fisher Scientific
AQ7100 AQ8100
76415-496EA 8643 CAD
76415-496 76415-498
Orion™ AquaMate Vis and UV-Vis Spectrophotometers, Thermo Scientific
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Orion™ AquaMate UV-Vis spectrophotometers include over 260 preprogrammed methods, and are designed to meet the needs of water and wastewater analysis laboratories.
  • Ideal for water and waste water analysis
  • With dual silicon photodiodes detector
  • Glove friendly touch-screen interface and keypad
AquaMate UV-Vis and Visible-only spectrophotometers incorporate advanced features and functions for ultimate flexibility in operation. Each verification report gives the time, date and instrument serial number in accordance with GLP and GMP.
More than 260More Product Information